Learn More
2-Fluoro-5-trifluoromethylbenzonitrile, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4088-84-0 |
| Molecular Formula | C8H3F4N |
| MDL Number | MFCD00061280 |
| Quantity | 10 g |
| SMILES | C1=C(C=CC(=C1C#N)F)C(F)(F)F |
| Molecular Weight (g/mol) | 189.11 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Fluoro-5-trifluoromethylbenzonitrile |