missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 11332679
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332679 1 g 1g
15427615 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332679 Supplier Thermo Scientific Alfa Aesar Supplier No. H26714.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As intermediates. Commonly used laboratory reagent. Used in the field of nanoparticles.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-98-9
Molecular Formula C9H6F4O3
Molecular Weight (g/mol) 238.14
MDL Number MFCD04115892
InChI Key PYCWSPHSXHTRQT-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethoxy phenylacetic acid, 2-2-fluoro-5-trifluoromethoxy phenyl acetic acid, 2-fluoro-5-trifluoromethoxy phenyl acetic acid
PubChem CID 2783328
IUPAC Name 2-[2-fluoro-5-(trifluoromethoxy)phenyl]acetic acid
SMILES OC(=O)CC1=C(F)C=CC(OC(F)(F)F)=C1

Specifications

Melting Point 54°C to 56°C
Quantity 1 g
Formula Weight 238.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)phenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.