missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-4-(methylsulfonyl)benzeneboronic acid, 98%

Product Code. 15420576
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420576 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420576 Supplier Alfa Aesar™ Supplier No. H53306.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 957060-85-4
Molecular Formula C7H8BFO4S
MDL Number MFCD09027248
Quantity 250 mg
Synonym 2-fluoro-4-methylsulfonyl phenylboronic acid, 2-fluoro-4-methylsulphonyl benzeneboronic acid, 2-fluoro-4-methylsulfonyl phenyl boronic acid, 2-fluoro-4-methanesulfonylphenylboronic acid, 2-fluoro-4-methanesulfonylphenyl boronic acid, 2-fluoro-4-methylsulfonyl benzeneboronic acid, boronic acid, b-2-fluoro-4-methylsulfonyl phenyl, 2-fluoro-4-methylsulfonyl phenylboronicacid, 2-fluoro-4-methylsulfonyl-phenylboronic acid
InChI Key VTSHDQJNJIIHGS-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC(F)=C(C=C1)B(O)O
IUPAC Name (2-fluoro-4-methylsulfonylphenyl)boronic acid
Molecular Weight (g/mol) 218.01
PubChem CID 44717666
Formula Weight 218.01
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-(methylsulfonyl)benzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.