missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 10389297 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10389297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10389297 Supplier Thermo Scientific Acros Supplier No. 430410010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 157834-21-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 60.9°C to 62.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H5BF4O2
MDL Number MFCD07784336
Quantity 1 g
Synonym 2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron, 2-fluoro-3-trifluoromethyl phenylboronicacid, boronic acid, 2-fluoro-3-trifluoromethyl phenyl, boronic acid,b-2-fluoro-3-trifluoromethyl phenyl, boronic acid, b-2-fluoro-3-trifluoromethyl phenyl
InChI Key PKARAEQMGHJHMY-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.92
PubChem CID 16217169
Formula Weight 207.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-3-(trifluoromethyl)phenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.