missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. p-7035645
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450626 1 g 1g
15460626 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450626 Supplier Thermo Scientific Alfa Aesar Supplier No. H53364.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-3-(trifluoromethyl)benzeneboronic acid is involved in suzuki-Miyaura coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157834-21-4
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.919
MDL Number MFCD07784336
InChI Key PKARAEQMGHJHMY-UHFFFAOYSA-N
Synonym 2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron, 2-fluoro-3-trifluoromethyl phenylboronicacid, boronic acid, 2-fluoro-3-trifluoromethyl phenyl, boronic acid,b-2-fluoro-3-trifluoromethyl phenyl, boronic acid, b-2-fluoro-3-trifluoromethyl phenyl
PubChem CID 16217169
IUPAC Name [2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 207.92
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.