missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylpyridine, 97%, Thermo Scientific™

Product Code. 11362946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362946 5 g 5g
11352946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362946 Supplier Thermo Scientific Alfa Aesar Supplier No. L10701.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2369-18-8
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041223
InChI Key PHGXUFPYCAWEHK-UHFFFAOYSA-N
Synonym 2-fluoro-3-picoline, 2-fluoro-3-methyl pyridine, 2-fluoro-3-methyl-pyridine, pyridine, 2-fluoro-3-methyl, 2-fluor-3-methylpyridin, pubchem3992, acmc-1ccv4, ksc497i8p, 2-fluoro-3-methylpyridine
PubChem CID 242722
IUPAC Name 2-fluoro-3-methylpyridine
SMILES CC1=C(N=CC=C1)F

Tekniske data

Density 1.078
Boiling Point 148°C to 150°C
Flash Point >100°C (212°F)
Refractive Index 1.475
Quantity 5 g
Beilstein 1877
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methylpyridine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.