missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-methylpyridine, 97%, Thermo Scientific™

Product Code. 11352946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352946 1 g 1g
11362946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352946 Supplier Thermo Scientific Alfa Aesar Supplier No. L10701.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2369-18-8
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041223
InChI Key PHGXUFPYCAWEHK-UHFFFAOYSA-N
Synonym 2-fluoro-3-picoline, 2-fluoro-3-methyl pyridine, 2-fluoro-3-methyl-pyridine, pyridine, 2-fluoro-3-methyl, 2-fluor-3-methylpyridin, pubchem3992, acmc-1ccv4, ksc497i8p, 2-fluoro-3-methylpyridine
PubChem CID 242722
IUPAC Name 2-fluoro-3-methylpyridine
SMILES CC1=C(N=CC=C1)F

Specifications

Density 1.078
Boiling Point 148°C to 150°C
Flash Point >100°C (212°F)
Refractive Index 1.475
Quantity 1 g
Beilstein 1877
Formula Weight 111.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.