Learn More
2-Fluoro-1,4-dimethoxybenzene, 97%, Thermo Scientific Chemicals
Product Code. 11383107
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383107 5 g 5g
11373107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11383107 Supplier Thermo Scientific Alfa Aesar Supplier No. L13505.06
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82830-49-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042487
InChI Key WNCYZVMZKSOPMU-UHFFFAOYSA-N
Synonym 1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
PubChem CID 145002
IUPAC Name 2-fluoro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)F
Specifications
Melting Point 23°C to 26°C
Boiling Point 119°C to 121°C (40 mmHg)
Refractive Index 1.5045
Quantity 5 g
Beilstein 6387735
Solubility Information Insoluble in water.
Formula Weight 156.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-1,4-dimethoxybenzene