missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyric acid, MP Biomedicals™

Product Code. 11415421 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415421 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415421 Supplier MP Biomedicals Supplier No. 0215797280

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 88-09-5
Color Colorless
Molecular Formula C6H12O2
Quantity 100 g
Synonym 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid
InChI Key OXQGTIUCKGYOAA-UHFFFAOYSA-N
SMILES CCC(CC)C(=O)O
IUPAC Name 2-ethylbutanoic acid
Molecular Weight (g/mol) 116.16
PubChem CID 6915
Formula Weight 116.16
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.