Learn More
2-Ethyl-anthraquinone, TRC
Product Code. 30008108
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008108 1 g 1g
30005573 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008108 Supplier Toronto Research Chemicals Supplier No. TRCE8993701G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 84-51-5
Molecular Formula C16 H12 O2
Molecular Weight (g/mol) 236.27
Synonym Anthraquinone, 2-ethyl- (7CI,8CI), 2-Ethyl-9,10-anthracenedione, 2-EAQ, 2-Ethyl-9,10-anthraquinone, 2-Ethylanthracene-9,10-dione, 2-Ethylanthraquinone, Kayacure 2-EAQ, NSC 7216, β-Ethylanthraquinone
IUPAC Name 2-ethylanthracene-9,10-dione
SMILES CCc1ccc2C(=O)c3ccccc3C(=O)c2c1
Spezifikation
Quantity 1 g
InChI Formula InChI=1S/C16H12O2/c1-2-10-7-8-13-14(9-10)16(18)12-6-4-3-5-11(12)15(13)17/h3-9H,2H2,1H3
Formula Weight 236.0837
Chemical Name or Material 2-Ethylanthraquinone