Learn More
2-Ethyl-6-methylaniline, TRC
Product Code. 30381079
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381079 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30381079 Supplier Toronto Research Chemicals Supplier No. TRCE922060100G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethyl-6-methylaniline
CAS 24549-06-2
Molecular Formula C9 H13 N
Quantity 100 g
Synonym Benzenamine, 2-ethyl-6-methyl-, o-Toluidine, 6-ethyl- (7CI,8CI), 2-Ethyl-6-methylbenzenamine, (2-Ethyl-6-methylphenyl)amine, 2-Ethyl-6-methylaniline, 2-Methyl-6-ethylaniline, 6-Ethyl-2-methylaniline, 6-Ethyl-o-toluidine, C 25702
InChI Formula InChI=1S/C9H13N/c1-3-8-6-4-5-7(2)9(8)10/h4-6H,3,10H2,1-2H3
SMILES CCc1cccc(C)c1N
IUPAC Name 2-ethyl-6-methylaniline
Molecular Weight (g/mol) 135.21
Formula Weight 135.1048