Learn More
2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™
Product Code. p-2698265
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10603423 1 g 1g
10131132 10 g 10g
10767485 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10603423 Supplier Thermo Scientific Maybridge Supplier No. SB01365DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 88634-80-4
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00173727
InChI Key RLBMKAXUSJOVJI-UHFFFAOYSA-N
PubChem CID 2795884
IUPAC Name 2-ethyl-5-methyl-1H-imidazole-4-carbaldehyde
SMILES CCC1=NC(C=O)=C(C)N1
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-ethyl-4-methyl-1h-imidazole-5-carbaldehyde, 2-ethyl-5-methyl-3h-imidazole-4-carbaldehyde, 1h-imidazole-5-carboxaldehyde,2-ethyl-4-methyl, 1h-imidazole-5-carboxaldehyde, 2-ethyl-4-methyl, 2-ethyl-5-formyl-4-methylimidazole, 2-ethyl-4-methylimidazole-5-carbaldehyde, 2-ethyl-4-methyl-1h-imidazole-5-carboxaldehyde, acmc-20lc80, 2-ethyl-4-methyl-5-formylimidazole, 1h-imidazole-4-carboxaldehyde, 2-ethyl-5-methyl
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
Product Title
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.