Learn More
2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers
Product Code. 11327179
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327179 5 g 5g
11337179 25 g 25g
11347179 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11327179 Supplier Thermo Scientific Alfa Aesar Supplier No. H27383.06
Request Bulk or Custom Format
CAS: 13925-07-0 | C8H12N2 | 136.198 g/mol
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13925-07-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00047392
InChI Key JZBCTZLGKSYRSF-UHFFFAOYSA-N
Synonym 3,5-dimethyl-2-ethylpyrazine, pyrazine, 2-ethyl-3,5-dimethyl, 2,6-dimethyl-3-ethylpyrazine, 3-ethyl-2,6-dimethylpyrazine, 2-ethyl-3,5-dimethyl pyrazine, pyrazine, 2,6-dimethyl-3-ethyl, unii-r182ey5l8c, fema no. 3149, 2-ethyl-3,5 6-dimethylpyrazine, pyrazine, 3,5-dimethyl-2-ethyl
PubChem CID 26334
IUPAC Name 2-ethyl-3,5-dimethylpyrazine
SMILES CCC1=NC=C(N=C1C)C
Specifications
Density 0.96
Boiling Point 182°C
Flash Point 66°C (151°F)
Refractive Index 1.5015
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,chloroform and dichloromethane.
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers
compliance-icons
Product Identifier
  • 2-Ethyl-3,5(6)-dimethylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-ethyl-3,5-dimethyl-pyrazine