Learn More
2-Ethyl-1,3-hexanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-1,3-hexandiol |
| CAS | 94-96-2 |
| Molecular Formula | C8 H18 O2 |
| Quantity | 100 g |
| Synonym | 1,3-Hexanediol, 2-ethyl-, 2-Ethyl-1,3-hexanediol, 2-Ethyl-1,3-hexylene glycol, 2-Ethyl-3-propyl-1,3-propanediol, 2-Ethylhexanediol, 3-Hydroxymethyl-n-heptan-4-ol, 6-12, Diol-Kyowa 8, Ethohexadiol, NSC 3881, Octylene glycol, Repellent 612, Rutgers 612 |
| InChI Formula | InChI=1S/C8H18O2/c1-3-5-8(10)7(4-2)6-9/h7-10H,3-6H2,1-2H3 |
| SMILES | CCCC(O)C(CC)CO |
| IUPAC Name | 2-ethylhexane-1,3-diol |
| Molecular Weight (g/mol) | 146.23 |
| Formula Weight | 146.1307 |