Learn More
2'-Ethoxyacetophenone, 97+%, Thermo Scientific Chemicals
Product Code. 11450631
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450631 50 g 50g
11440631 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11450631 Supplier Thermo Scientific Alfa Aesar Supplier No. B20113.18
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 2142-67-8 | Molecular Formula: C10H12O2 | Molecular Weight: 164.204 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2142-67-8
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00017262
InChI Key TVGMOUGXQYQZOL-UHFFFAOYSA-N
Synonym 2'-ethoxyacetophenone, 1-2-ethoxyphenyl ethanone, ethanone, 1-2-ethoxyphenyl, 2-acetylphenetole, 1-2-ethoxyphenyl ethan-1-one, 1-acetyl-2-ethoxybenzene, o-ethoxyacetophenone, pubchem13191, acmc-1bt18, ethanone,2-ethoxy-1-phenyl
PubChem CID 578482
IUPAC Name 1-(2-ethoxyphenyl)ethanone
SMILES CCOC1=CC=CC=C1C(=O)C
Specifications
Melting Point 38°C to 42°C
Density 1
Boiling Point 244°C
Flash Point >110°C (230°F)
Quantity 50 g
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 2'-Ethoxyacetophenone