Learn More
2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane |
| CAS | 13679-86-2 |
| Molecular Formula | C10H16O |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H16O/c1-5-10(4)7-6-9(11-10)8(2)3/h5,9H,1-2,6-7H2,3-4H3 |
| SMILES | CC(=C)C1CCC(C)(O1)C=C |
| IUPAC Name | 2-ethenyl-2-methyl-5-prop-1-en-2-yloxolane |
| Molecular Weight (g/mol) | 152.23 |
| Formula Weight | 152.1201 |
Product Title
Vous avez repéré une opportunité d'amélioration ?