missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)benzaldehyde oxime, 95%, Thermo Scientific Chemicals

Product Code. 15418397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418397 1 g 1g
15428397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418397 Supplier Thermo Scientific Alfa Aesar Supplier No. H51903.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153358-05-5 | Molecular Formula: C19H16NOP | Molecular Weight: 305.32 g/mol

It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis, structure and applications to the catalytic rearrangement and dehydration of aldoximes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153358-05-5
Molecular Formula C19H16NOP
Molecular Weight (g/mol) 305.32
MDL Number MFCD16251585
InChI Key XHIVESUSSLEMGJ-UHFFFAOYSA-N
Synonym 2-diphenylphosphino benzaldehyde oxime, n-2-diphenylphosphino benzylidene hydroxylamine, e-n-2-diphenylphosphanyl phenyl methylidene hydroxylamine
PubChem CID 98043674
IUPAC Name (NZ)-N-[(2-diphenylphosphanylphenyl)methylidene]hydroxylamine
SMILES ON=CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 305.32
Percent Purity 95%
Chemical Name or Material 2-(Diphenylphosphino)benzaldehyde oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.