Learn More
2-Diisopropylaminoethyl Ester Xanthene-9-carboxylic Acid Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Diisopropylaminoethyl Ester Xanthene-9-carboxylic Acid Hydrochloride |
| CAS | 102476-84-6 |
| Molecular Formula | C22H27NO3.(HCl) |
| Quantity | 10 mg |
| Synonym | 2-[Bis(1-methylethyl)amino]ethyl Ester 9H-Xanthene-9-carboxylic Acid Hydrochloride, 2-(Diisopropylamino)ethyl 9H-Xanthene-9-carboxylate Hydrochloride |
| InChI Formula | InChI=1S/C22H27NO3.ClH/c1-15(2)23(16(3)4)13-14-25-22(24)21-17-9-5-7-11-19(17)26-20-12-8-6-10-18(20)21;/h5-12,15-16,21H,13-14H2,1-4H3;1H |
| SMILES | Cl.CC(C)N(CCOC(=O)C1c2ccccc2Oc3ccccc13)C(C)C |
| IUPAC Name | 2-[di(propan-2-yl)amino]ethyl 9H-xanthene-9-carboxylate;hydrochloride |
| Molecular Weight (g/mol) | 389.9 |
| Formula Weight | 389.1758 |