missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine Monohydrate, MP Biomedicals™

Product Code. 11483440 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483440 5 g 5g
11493440 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483440 Supplier MP Biomedicals Supplier No. 0210148105
  • ONLINE EXCLUSIVE PRICE
    326.00 EUR
    309.75 EUR / 5g
    Save 16.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 16373-93-6
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00149364
InChI Key OLXZPDWKRNYJJZ-YUZWJPFSNA-N
Synonym 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
PubChem CID 9549172
IUPAC Name (2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White
Assay Percent Range 98+%
Quantity 5 g
Formula Weight 269.2
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine Monohydrate, FTIR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.