missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals™

Product Code. 11462192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11462192 Supplier MP Biomedicals Supplier No. 0215079410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-Deoxyadenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals may be used for the synthesis of deoxyadenylate oligonucleotides with enzymes such as polynucleotide phosphorylase from Escherichia coli.

dADP is a bacterial poly(A)polymerase inhibitor that may be used as an alternative substrate or inhibitor for studies of ATPase and DNA and RNA polymerase specificity.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72003-83-9
Molecular Formula C10H15N5Na2O9P2
MDL Number MFCD00083610,MFCD00083610
Quantity 10 mg
Synonym 2'-deoxyadenosine-5'-diphosphate disodium salt
InChI Key PMCITFSCGIPAKY-UHFFFAOYNA-N
SMILES [Na].[Na].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP(O)(O)=O)O3)C2=NC=N1
IUPAC Name [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid disodium
Molecular Weight (g/mol) 457.18
PubChem CID 131858105
Formula Weight 397.2
Percent Purity ≥97%
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.