missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine-5'-diphosphate Trisodium Salt, ≥97%, MP Biomedicals™

Product Code. 11472192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472192 25 mg 25mg
11482192 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472192 Supplier MP Biomedicals Supplier No. 0215079425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 72003-83-9
Molecular Formula C10H15N5Na2O9P2
Molecular Weight (g/mol) 457.18
MDL Number MFCD00083610,MFCD00083610
InChI Key PMCITFSCGIPAKY-UHFFFAOYNA-N
Synonym 2'-deoxyadenosine-5'-diphosphate disodium salt
PubChem CID 131858105
IUPAC Name [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid disodium
SMILES [Na].[Na].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP(O)(O)=O)O3)C2=NC=N1

Specifications

Color White
Assay Percent Range ≥97%
Quantity 25 mg
Formula Weight 397.2
Physical Form Amorphous Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-diphosphate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.