Learn More
2'-Deoxyadenosine-3'-monophosphate xSodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 2'-Deoxyadenosine-3'-monophosphate xSodium Salt |
| CAS | 102832-10-0 |
| Molecular Formula | C10H14N5O6P • xNa |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C10H14N5O6P.Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(6(2-16)20-7)21-22(17,18)19;/h3-7,16H,1-2H2,(H2,11,12,13)(H2,17,18,19);/t5-,6+,7+;/m0./s1 |
| SMILES | NC(N=CN=C12)=C1N=CN2[C@@H]3O[C@H](CO)[C@@H](OP(O)(O)=O)C3.[Na] |
| IUPAC Name | (2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-yl dihydrogen phosphate, sodium salt |
| Molecular Weight (g/mol) | 354.21 |
| Formula Weight | 354.06 |