Learn More
2'-Deoxy-N3-methyluridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2'-Deoxy-N3-methyluridine |
| CAS | 24514-32-7 |
| Molecular Formula | C10H14N2O5 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C10H14N2O5/c1-11-8(15)2-3-12(10(11)16)9-4-6(14)7(5-13)17-9/h2-3,6-7,9,13-14H,4-5H2,1H3/t6-,7+,9+/m0/s1 |
| SMILES | CN1C(=O)C=CN(C1=O)[C@H]2C[C@@H]([C@H](O2)CO)O |
| Molecular Weight (g/mol) | 242.23 |
| Formula Weight | 242.0903 |