missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxy-3’,5’-di-O-p-toluoyl Uridine-13C,15N2, TRC

Product Code. 30355560
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355560 5 mg 5mg
30355573 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355560 Supplier Toronto Research Chemicals Supplier No. TRCD2328975MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2’-Deoxy-3’,5’-di-O-p-toluoyl Uridine-13C,15N2
Molecular Formula 13C C24 H24 15N2 O7
Quantity 5 mg
Synonym 2'-Deoxyuridine 3',5'-Bis(4-methylbenzoate), 2'-Deoxyuridine 3',5'-Di-p-toluate
InChI Formula InChI=1S/C25H24N2O7/c1-15-3-7-17(8-4-15)23(29)32-14-20-19(34-24(30)18-9-5-16(2)6-10-18)13-22(33-20)27-12-11-21(28)26-25(27)31/h3-12,19-20,22H,13-14H2,1-2H3,(H,26,28,31)/t19?,20-,22-/m1/s1/i25+1,26+1,27+1
SMILES Cc1ccc(cc1)C(=O)OC[C@H]2O[C@H](CC2OC(=O)c3ccc(C)cc3)[15N]4C=CC(=O)[15NH][13C]4=O
IUPAC Name [(2R,5R)-5-(2,4-dioxo(213C,1,3-15N2)pyrimidin-1-yl)-3-(4-methylbenzoyl)oxyoxolan-2-yl]methyl 4-methylbenzoate
Molecular Weight (g/mol) 467.447
Formula Weight 467.156
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.