Learn More
2-(Cyclopropylmethoxy)-acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Cyclopropylmethoxy)-acetic Acid |
| CAS | 246869-08-9 |
| Molecular Formula | C6H10O3 |
| Quantity | 250 mg |
| Synonym | (Cyclopropylmethoxy)acetic Acid |
| InChI Formula | InChI=1S/C6H10O3/c7-6(8)4-9-3-5-1-2-5/h5H,1-4H2,(H,7,8) |
| SMILES | OC(=O)COCC1CC1 |
| IUPAC Name | 2-(cyclopropylmethoxy)acetic acid |
| Molecular Weight (g/mol) | 130.14 |
| Formula Weight | 130.063 |