Learn More
2-(Cyclopropylmethoxy)-acetic Acid 1,1-Dimethyl-2-[4-(methylsulfonyl)phenyl]-2-oxoethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Cyclopropylmethoxy)-acetic Acid 1,1-Dimethyl-2-[4-(methylsulfonyl)phenyl]-2-oxoethyl Ester |
| CAS | 246869-15-8 |
| Molecular Formula | C17H22O6S |
| Quantity | 1 mg |
| Synonym | 2-Methyl-1-[4-(methylsulfonyl)phenyl]-1-oxo-2-propanyl (cyclopropylmethoxy)acetate |
| InChI Formula | InChI=1S/C17H22O6S/c1-17(2,23-15(18)11-22-10-12-4-5-12)16(19)13-6-8-14(9-7-13)24(3,20)21/h6-9,12H,4-5,10-11H2,1-3H3 |
| SMILES | CC(C)(OC(=O)COCC1CC1)C(=O)c2ccc(cc2)S(=O)(=O)C |
| IUPAC Name | [2-methyl-1-(4-methylsulfonylphenyl)-1-oxopropan-2-yl] 2-(cyclopropylmethoxy)acetate |
| Molecular Weight (g/mol) | 354.42 |
| Formula Weight | 354.1137 |