Learn More
2-Cyanomethyl-4-(4-hydroxyphenyl)thiazole, 97%, Thermo Scientific™
Product Code. 15484528
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484528 2 g 2g
15494528 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15484528 Supplier Thermo Scientific Alfa Aesar Supplier No. H58206.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1028352-86-4
Molecular Formula C11H8N2OS
Molecular Weight (g/mol) 216.258
MDL Number MFCD22682948
InChI Key CMIGLFUTBISGQV-UHFFFAOYSA-N
Synonym 2-cyanomethyl-4-4-hydroxyphenyl thiazole, 2-4-4-hydroxyphenyl-1,3-thiazol-2-yl acetonitrile
PubChem CID 73996315
IUPAC Name 2-[4-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetonitrile
SMILES C1=CC(=CC=C1C2=CSC(=N2)CC#N)O
Specifications
Melting Point 145°C to 151°C
Quantity 2 g
UN Number UN3439
Formula Weight 216.25
Percent Purity 97%
Chemical Name or Material 2-Cyanomethyl-4-(4-hydroxyphenyl)thiazole