missing translation for 'onlineSavingsMsg'
Learn More

2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%

Product Code. p-3211670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10568093 1 g Glass bottle 1g
10302103 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10568093

Brand: Thermo Scientific Acros 374830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89992-70-1 | C9H18ClN2OP | 236.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89992-70-1
Molecular Formula C9H18ClN2OP
Molecular Weight (g/mol) 236.68
MDL Number MFCD00011544
InChI Key QWTBDIBOOIAZEF-UHFFFAOYSA-N
Synonym 2-cyanoethyl n,n-diisopropylchlorophosphoramidite, 2-cyanoethyl-n,n-diisopropylchlorophosphoramidite, phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethoxy diisopropylamino chlorophosphine, 3-chloro diisopropylamino phosphino oxy propanenitrile, 3-chloro-diisopropylamino phosphanyl oxypropanenitrile, 3-chloro diisopropylamino phosphanyl oxy propanenitrile, acmc-20anxd, 2-cyanoethyl diisopropylchloro-phosphoramidite, 2-cyanoethyl diisopropylphosphoramidochloridite
PubChem CID 2734844
IUPAC Name 3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile
SMILES CC(C)N(C(C)C)P(OCCC#N)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Density 1.0610g/mL
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(CH3)2CH]2NP(Cl)O(CH2)2CN
Refractive Index 1.4760 to 1.4800
Quantity 1 g
Specific Gravity 1.061
Solubility Information Solubility in water: reacts.
Formula Weight 236.68
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H250-Catches fire spontaneously if exposed to air.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.