accessibility menu, dialog, popup

UserName

H3CCH3NCH3H3CPONCl

CAS RN 89992-70-1

IUPAC Name: 3-({[bis(propan-2-yl)amino](chloro)phosphanyl}oxy)propanenitrile
Synonyms: 3-{[chloro(diisopropylamino)phosphanyl]oxy}propanenitrile
Molecular Weight (g/mol): 236.68
Molecular Formula: C9H18ClN2OP
InChi Key: QWTBDIBOOIAZEF-UHFFFAOYNA-N
SMILES: CC(C)N(C(C)C)P(Cl)OCCC#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)