Learn More
2-Cyanobenzaldehyde, TRC
Product Code. 30360488
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30360488 Supplier Toronto Research Chemicals Supplier No. TRCC9622705G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7468-67-9
Molecular Formula C8H5NO
Quantity 5 g
Synonym 2-Formylbenzonitrile, o-Cyanobenzaldehyde, Phthalaldehydonitrile
InChI Formula InChI=1S/C8H5NO/c9-5-7-3-1-2-4-8(7)6-10/h1-4,6H
SMILES O=Cc1ccccc1C#N
IUPAC Name 2-formylbenzonitrile
Molecular Weight (g/mol) 131.13
Formula Weight 131.0371
Chemical Name or Material 2-Cyanobenzaldehyde