Learn More
2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, TRC
Product Code. 30135721
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135721 50 g 50g
30133725 10 g 10g
30011904 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135721 Supplier Toronto Research Chemicals Supplier No. TRCC98224550G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24522-30-3
Molecular Formula C10 H7 F3 N2 O
Molecular Weight (g/mol) 228.17
Synonym 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide, Leflunomide Imp. H (EP)
IUPAC Name 2-cyano-N-[4-(trifluoromethyl)phenyl]acetamide
SMILES FC(F)(F)c1ccc(NC(=O)CC#N)cc1
Specifikationer
Quantity 50 g
InChI Formula InChI=1S/C10H7F3N2O/c11-10(12,13)7-1-3-8(4-2-7)15-9(16)5-6-14/h1-4H,5H2,(H,15,16)
Formula Weight 228.051
Chemical Name or Material 2-Cyano-N-[4-(trifluoromethyl)phenyl]acetamide
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.