Learn More
2-Cyano Loratadine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Cyano Loratadine |
| CAS | 860010-31-7 |
| Molecular Formula | C23 H22 Cl N3 O2 |
| Quantity | 1 mg |
| Synonym | 1-Piperidinecarboxylic acid, 4-(8-chloro-2-cyano-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-, ethyl ester, 2-Cyano Loratadine, 2-Cyanoloratadine, Ethyl 4-(8-chloro-2-cyano-5,6-dihydrobenzo[1,2]cyclohepta[2,4-c]pyridin-11-ylidene)piperidine-1-carboxylate, Ethyl 4-(8-chloro-2-cyano-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate |
| InChI Formula | InChI=1S/C23H22ClN3O2/c1-2-29-23(28)27-11-9-15(10-12-27)21-20-8-6-18(24)13-17(20)4-3-16-5-7-19(14-25)26-22(16)21/h5-8,13H,2-4,9-12H2,1H3 |
| SMILES | CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4ccc(nc24)C#N)CC1 |
| IUPAC Name | ethyl 4-(8-chloro-2-cyano-5,6-dihydrobenzo[1,2]cyclohepta[3,4-c]pyridin-11-ylidene)piperidine-1-carboxylate |
| Molecular Weight (g/mol) | 407.89 |
| Formula Weight | 407.1401 |