Learn More
2-Chloropyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific™
Product Code. 11414480
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414480 1 g 1g
11424480 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11414480 Supplier Thermo Scientific Alfa Aesar Supplier No. H50070.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 458532-84-8
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.506
MDL Number MFCD04039870
InChI Key UUEQDBHKMOFLDP-UHFFFAOYSA-N
Synonym 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridine-4-boronic acid pinacol ester, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloropyridin-4-ylboronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloropyridine-4-boronic acid, pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16684
PubChem CID 5103408
IUPAC Name 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Cl
Specifications
Quantity 1 g
Formula Weight 239.51
Percent Purity 98%
Chemical Name or Material 2-Chloropyridine-4-boronic acid pinacol ester