Learn More
2-Chloropropene, 99%
Product Code. 11358775
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358775 25 g 25g
11348775 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358775 Supplier Thermo Scientific Alfa Aesar Supplier No. L12374.14
 Restricted Item
In Stock
Add to basket
CAS: 557-98-2 | C3H5Cl | 76.523 g/mol
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 557-98-2
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.523
MDL Number MFCD00000859
InChI Key PNLQPWWBHXMFCA-UHFFFAOYSA-N
Synonym 2-chloropropene, 1-propene, 2-chloro, isopropenyl chloride, 2-chloropropylene, 2-chloro-1-propene, beta-chloropropene, beta-chloropropylene, propene, 2-chloro, unii-50txn55e4a, 2-chloro propylene
PubChem CID 11203
IUPAC Name 2-chloroprop-1-ene
SMILES CC(=C)Cl
Specifications
Melting Point -138°C
Density 0.899
Boiling Point 22°C to 23°C
Flash Point −34°C (−29°F)
Odor Recognizable
Refractive Index 1.3939
Quantity 25 g
UN Number UN2456
Beilstein 1361376
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 76.53
Percent Purity 99%
Chemical Name or Material 2-Chloropropene