Learn More
2-Chloropropane, TRC
Artikelnummer. 30349785
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity
30349785 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30349785 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC37920550ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material 2-Chloropropane (Isopropylchloride)
CAS 75-29-6
Molecular Formula C3 H7 Cl
Quantity 50 mL
Synonym Propane, 2-chloro- (8CI, 9CI, ACI), 2-Chloropropane (ACI), 2-Propyl chloride, Chlorodimethylmethane, Isoprid, Isopropyl chloride, LBL 2, Narcosop, R 280da, sec-Propyl chloride
InChI Formula InChI=1S/C3H7Cl/c1-3(2)4/h3H,1-2H3
SMILES CC(C)Cl
IUPAC Name 2-chloropropane
Molecular Weight (g/mol) 78.54
Formula Weight 78.0236