Learn More
2-Chloropropane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloropropane (Isopropylchloride) |
| CAS | 75-29-6 |
| Molecular Formula | C3 H7 Cl |
| Quantity | 25 mL |
| Synonym | Propane, 2-chloro- (8CI, 9CI, ACI), 2-Chloropropane (ACI), 2-Propyl chloride, Chlorodimethylmethane, Isoprid, Isopropyl chloride, LBL 2, Narcosop, R 280da, sec-Propyl chloride |
| InChI Formula | InChI=1S/C3H7Cl/c1-3(2)4/h3H,1-2H3 |
| SMILES | CC(C)Cl |
| IUPAC Name | 2-chloropropane |
| Molecular Weight (g/mol) | 78.54 |
| Formula Weight | 78.0236 |