missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropane, Sigma-Aldrich™

Product Code. 32237637
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237637 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237637 Supplier Sigma Aldrich Supplier No. 8028092500
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloropropane
CAS 75-29-6
Melting Point -118°C
Density 0.86 g/cm³ at 20°C
Flash Point -36°C
Molecular Formula (CH3)2CHCl
Quantity 2.5 L
Synonym 2-Chloropropane, Isopropyl chloride, 2-Chloropropane
Molecular Weight (g/mol) 78.54
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.