Learn More
2-Chlorophenylacetonitrile, 98+%
Product Code. 11424574
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424574 10 g 10g
11434574 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11424574 Supplier Thermo Scientific Alfa Aesar Supplier No. A13176.09
 Restricted Item
In Stock
Add to basket
CAS: 2856-63-5 | C8H6ClN | 151.593 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2856-63-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001898
InChI Key MRDUURPIPLIGQX-UHFFFAOYSA-N
Synonym 2-chlorobenzyl cyanide, 2-chlorophenylacetonitrile, 2-2-chlorophenyl acetonitrile, o-chlorobenzyl cyanide, benzeneacetonitrile, 2-chloro, 2-chlorobenzeneacetonitrile, 2-chlorobenzylcyanide, 2-chlorophenyl acetonitrile, o-chlorophenyl acetonitrile, acetonitrile, o-chlorophenyl
PubChem CID 76112
IUPAC Name 2-(2-chlorophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)Cl
Specifications
Melting Point 21°C to 24°C
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 10 g
UN Number UN3439
Beilstein 2043683
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenylacetonitrile