Learn More
2-Chloromethyl-1,4-benzodioxane, 97%, Thermo Scientific™
Product Code. 11366806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366806 25 g 25g
11356806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11366806 Supplier Thermo Scientific Alfa Aesar Supplier No. L04137.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2164-33-2
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00014666
InChI Key AYPKYQSHFKQVDL-UHFFFAOYSA-N
Synonym 2-chloromethyl-2,3-dihydro-1,4-benzodioxine, 2-chloromethyl-1,4-benzodioxane, 2-chloromethyl-2,3-dihydrobenzo b 1,4 dioxine, 2-chloromethyl-2,3-dihydro-1,4-benzodioxin, 2-chloromethyl-2,3-dihydro-benzo 1,4 dioxine, 2-chloromethyl-2h,3h-benzo e 1,4-dioxane, acmc-1cfd2, 2-chloromethyl-benzodioxane, 2-chloromethyl-1,4-benzodioxan
PubChem CID 95320
IUPAC Name 3-(chloromethyl)-2,3-dihydro-1,4-benzodioxine
SMILES C1C(OC2=CC=CC=C2O1)CCl
Specifications
Boiling Point 96°C (0.4 mmHg)
Refractive Index 1.554
Quantity 25 g
Beilstein 150153
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 2-Chloromethyl-1,4-benzodioxane