Learn More
2-Chlorobenzotrichloride, TRC
Specifications
Specifications
| Chemical Name or Material | alpha,alpha,alpha-2-Tetrachlorotoluene |
| CAS | 2136-89-2 |
| Molecular Formula | C7 H4 Cl4 |
| Quantity | 50 g |
| Synonym | (2-Chlorophenyl)trichlormethane, Toluene, o,α,α,α-tetrachloro- (6CI,7CI,8CI), 1-Chloro-2-(trichloromethyl)benzene, 2-Chlorobenzotrichloride, NSC 59736, o-Chlorobenzotrichloride, o-Chlorobenzylidyne chloride, o-Chlorophenyltrichloromethane, α,α,α,2-Tetrachlorotoluene |
| InChI Formula | InChI=1S/C7H4Cl4/c8-6-4-2-1-3-5(6)7(9,10)11/h1-4H |
| SMILES | Clc1ccccc1C(Cl)(Cl)Cl |
| IUPAC Name | 1-chloro-2-(trichloromethyl)benzene |
| Molecular Weight (g/mol) | 229.92 |
| Formula Weight | 227.9067 |