missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)aniline, 97%

Product Code. p-7026622
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445367 5 g 5g
11455367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445367 Supplier Thermo Scientific Alfa Aesar Supplier No. B22714.06
Add to basket
This item is not returnable. View return policy

CAS: 433-94-3 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-94-3
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00272565
InChI Key OTRRSPQJZRCMDA-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl aniline, 2-amino-3-chlorobenzotrifluoride, benzenamine, 2-chloro-6-trifluoromethyl, 2-chloranyl-6-trifluoromethyl aniline, pubchem2764, 2-chloro-6-trifluoromethyl benzenamine, 2-chloro-6-trifluoromethyl phenylamine, 6-chloro-2-trifluoromethyl phenylamine, 2-chloro-6-trifluoromethyl-benzenamine, 2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluoride
PubChem CID 2756384
IUPAC Name 2-chloro-6-(trifluoromethyl)aniline
SMILES NC1=C(Cl)C=CC=C1C(F)(F)F

Specifications

Density 1.42
Boiling Point 39°C to 40°C (0.1 mmHg)
Quantity 5 g
UN Number UN2810
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.