missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylisonicotinic acid, 95+%, Thermo Scientific™

Product Code. 10269763
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269763 25 g Amber Glass Bottle 25g
10510743 1 g Amber Glass Bottle 1g
10520743 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269763 Supplier Thermo Scientific Maybridge Supplier No. RF03759EB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25462-85-5
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
InChI Key ZGZMEKHQIZSZOH-UHFFFAOYSA-N
Synonym 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
PubChem CID 141209
IUPAC Name 2-chloro-6-methylpyridine-4-carboxylic acid
SMILES CC1=NC(=CC(=C1)C(=O)O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 171.58
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.