Learn More
2-Chloro-5-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
Product Code. p-7035285
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450286 250 mg 250mg
15440286 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15450286 Supplier Thermo Scientific Alfa Aesar Supplier No. H52999.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Boronic acids are often used in metal-catalyzed cross-coupling for C-C, C-N bond formation, and C-H functionalization. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates. Other known chemistry may includes 1,4-addition to activated alkenes, 1,2-addition to carbonyls and imines, addition to anhydrides, 1,4-addition to a,ß-unsaturated ketones(4), and 1,4-addition to esters and amides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 913835-92-4
Molecular Formula C8H8BClO4
Molecular Weight (g/mol) 214.408
MDL Number MFCD08689528
InChI Key LZMDHZSRUQKDFI-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxycarbonyl phenylboronic acid, methyl 3-borono-4-chlorobenzoate, 2-chloro-5-methoxycarbonyl phenyl boronic acid, 2-chloro-5-methoxycarbonyl phenylboronicacid, acmc-209rad, 2-chloro-5-methoxycarbonylphenyl boronic acid, 2-chloranyl-5-methoxycarbonyl-phenyl boronic acid, benzoicacid, 3-borono-4-chloro-, 1-methyl ester 9ci
PubChem CID 44119360
IUPAC Name (2-chloro-5-methoxycarbonylphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)OC)Cl)(O)O
Specifications
Melting Point 230°C to 232°C
Quantity 250 mg
Formula Weight 214.41
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid