accessibility menu, dialog, popup

UserName

CAS RN 913835-92-4

BOOH3CClHOOH

CAS RN 913835-92-4

IUPAC Name: [2-chloro-5-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 2-chloro-5-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 214.41
Molecular Formula: C8H8BClO4
InChi Key: LZMDHZSRUQKDFI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(B(O)O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results