Learn More
2-Chloro-5-methoxybenzimidazole, 94%
Product Code. p-7044845
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338175 250 mg 250mg
11328175 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338175 Supplier Thermo Scientific Alfa Aesar Supplier No. L08440.MD
Add to basket
CAS: 15965-54-5 | C8H7ClN2O | 182.607 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15965-54-5
Molecular Formula C8H7ClN2O
Molecular Weight (g/mol) 182.607
MDL Number MFCD00051945
InChI Key FMDGYQOERIOABX-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxybenzimidazole, 2-chloro-5-methoxy-1h-benzo d imidazole, 2-chloro-5-methoxy-1h-benzimidazole, 2-chloro-5-methoxy-1h-1,3-benzodiazole, 2-chloro-5-methoxy-3h-1,3-benzodiazole, acmc-1bs4u, 5-methoxy-2-chlorobenzimidazole, 2-chloro-5-methoxybenzoimidazole, 1h-benzimidazole,2-chloro-6-methoxy, 1h-benzimidazole, 2-chloro-5-methoxy
PubChem CID 519197
IUPAC Name 2-chloro-6-methoxy-1H-benzimidazole
SMILES COC1=CC2=C(C=C1)N=C(N2)Cl
Specifications
Melting Point 165°C to 168°C
Quantity 250 mg
Beilstein 777455
Formula Weight 182.61
Percent Purity 94%
Chemical Name or Material 2-Chloro-5-methoxybenzimidazole