Learn More
2-Chloro-5-(2H-tetrazol-5-yl)-4-((thiophen-2-ylmethyl)amino)benzenesulfonamide, 97%, BLD Pharm™
Product Code. 30948856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
25 g
Unit Size:
100mg
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948856 1 g 1g
30936788 10 g 10g
30944215 250 mg 250mg
30948264 25 g 25g
30934156 100 mg 100mg
30938218 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30948856 Supplier BLD Pharm Supplier No. BD1149841g
 Restricted Item
Estimated Shipment: 27-11-2026
Especificaciones
CAS 27589-33-9
Molecular Formula C12H11ClN6O2S2
MDL Number MFCD30541344
Quantity 1 g
SMILES O=S(C1=CC(C2=NNN=N2)=C(NCC3=CC=CS3)C=C1Cl)(N)=O
Molecular Weight (g/mol) 370.84
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(2H-tetrazol-5-yl)-4-((thiophen-2-ylmethyl)amino)benzenesulfonamide