missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 15496187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15496187 1 g 1g
15438435 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15496187

Brand: Thermo Scientific Alfa Aesar H33376.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56456-51-0
Molecular Formula C8H6ClF3O
Molecular Weight (g/mol) 210.58
MDL Number MFCD04972757
InChI Key GMZPPTCATYZORA-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzyl alcohol, 2-chloro-4-trifluoromethyl benzylalcohol, 2-chloro-4-trifluoromethylbenzyl alcohol, 2-chloro-4-trifluoromethyl phenyl methanol, 2-chlor-4-trifluoromethyl-benzylalcohol, benzenemethanol,2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethylbenzylalcohol, 2-chloro-4 trifluoromethyl benzyl alcohol
PubChem CID 40427110
IUPAC Name [2-chloro-4-(trifluoromethyl)phenyl]methanol
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)CO

Specifications

Melting Point 50°C to 52°C
Quantity 1 g
Formula Weight 210.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.