accessibility menu, dialog, popup

UserName

CAS RN 56456-51-0

FFFClOH

CAS RN 56456-51-0

IUPAC Name: [2-chloro-4-(trifluoromethyl)phenyl]methanol
Synonyms: [2-chloro-4-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 210.58
Molecular Formula: C8H6ClF3O
InChi Key: GMZPPTCATYZORA-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results