Learn More
2-Chloro-4-nitrophenyl 2-Acetamido-2-deoxy-b-d-glucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4-nitrophenyl 2-Acetamido-2-deoxy-b-d-glucopyranoside |
| CAS | 103614-82-0 |
| Molecular Formula | C14H17N2O8Cl |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C14H17ClN2O8/c1-6(19)16-11-13(21)12(20)10(5-18)25-14(11)24-9-3-2-7(17(22)23)4-8(9)15/h2-4,10-14,18,20-21H,5H2,1H3,(H,16,19)/t10-,11-,12-,13-,14-/m1/s1 |
| SMILES | CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2ccc(cc2Cl)[N+](=O)[O-] |
| IUPAC Name | N-[(2S,3R,4R,5S,6R)-2-(2-chloro-4-nitrophenoxy)-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| Molecular Weight (g/mol) | 376.75 |
| Formula Weight | 376.0673 |