missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorocinnamic acid, 97%, Thermo Scientific™

Product Code. 11416256
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416256 25 g 25g
11496246 1 g 1g
11406256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416256 Supplier Thermo Scientific Alfa Aesar Supplier No. B23062.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133220-86-7
Molecular Formula C9H6ClFO2
Molecular Weight (g/mol) 200.593
MDL Number MFCD00236027
InChI Key RJCWBTRMWGOREZ-DUXPYHPUSA-N
Synonym 2-chloro-4-fluorocinnamic acid, rarechem bk hw 0063, timtec-bb sbb003541, 3-2-chloro-4-fluorophenyl-2-propenoic acid, 2e-3-2-chloro-4-fluorophenyl prop-2-enoic acid, 3-2-chloro-4-fluorophenyl acrylic acid, pubchem3429, e-3-2-chloro-4-fluorophenyl acrylic acid, 3-chloro-5-fluorobenzoic, 2-chloro-4-fluorocinnamicacid
PubChem CID 6257297
IUPAC Name (E)-3-(2-chloro-4-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1F)Cl)C=CC(=O)O

Specifications

Melting Point 245°C to 249°C
Quantity 25 g
Formula Weight 200.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorocinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.